Assay Details
Target Gene Details
Entrez Gene ID: | 91351 |
Gene Name: | DEAD-box helicase 60-like |
Gene Aliases: |
- |
Location: |
Chr.4:168356735-168480514 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 44 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX60L | NM_001012967.2 | NP_001012985.2 | ||
XM_005263341.4 | XP_005263398.2 | |||
XM_005263342.2 | XP_005263399.2 | |||
XM_005263343.2 | XP_005263400.2 | |||
XM_005263345.4 | XP_005263402.2 | |||
XM_006714408.2 | XP_006714471.1 | |||
XM_011532402.2 | XP_011530704.1 | |||
XM_011532403.2 | XP_011530705.1 | |||
XM_011532404.1 | XP_011530706.1 | |||
XM_011532405.1 | XP_011530707.1 | |||
XM_011532406.1 | XP_011530708.1 | |||
XM_011532409.2 | XP_011530711.1 | |||
XM_017008829.1 | XP_016864318.1 | |||
XM_017008830.1 | XP_016864319.1 | |||
XM_017008831.1 | XP_016864320.1 | |||
AK023530.1 | ||||
AK023743.1 | ||||
AL832950.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |