Assay Details
Target Gene Details
Entrez Gene ID: | 285527 |
Gene Name: | FRY like transcription coactivator |
Gene Aliases: |
AF4p12, KIAA0826, MOR2 |
Location: |
Chr.4:48497363-48780312 on Build GRCh38 |
Assay Gene Location: | Within Intron 58 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FRYL | NM_015030.1 | NP_055845.1 | ||
XM_005248090.4 | XP_005248147.1 | |||
XM_005248093.4 | XP_005248150.1 | |||
XM_011513680.2 | XP_011511982.1 | |||
XM_011513681.2 | XP_011511983.1 | |||
XM_011513682.2 | XP_011511984.1 | |||
XM_011513683.2 | XP_011511985.1 | |||
XM_011513684.2 | XP_011511986.1 | |||
XM_017008046.1 | XP_016863535.1 | |||
AB020633.1 | BAA74849.1 | |||
AK127307.1 | ||||
AL833170.1 | ||||
BC021803.1 | AAH21803.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829925 | Chr.4:48465676 - 48637325 on Build GRCh38 | Gain+Loss | ZAR1 SLC10A4 FRYL |
nsv594121 | Chr.4:48425756 - 48603611 on Build GRCh38 | Gain | ZAR1 SLC10A4 SLAIN2 FRYL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |