Assay Details
Target Gene Details
Entrez Gene ID: | 100130872 |
Gene Name: | uncharacterized LOC100130872 |
Gene Aliases: |
- |
Location: |
Chr.4:1195783-1208962 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100130872 | NR_024569.1 | 3 | 580 | |
AK091779.1 | BAC03745.1 | |||
CR627052.1 | 3 | 580 |
Target Gene Details
Entrez Gene ID: | 10417 |
Gene Name: | spondin 2 |
Gene Aliases: |
DIL-1, DIL1, M-SPONDIN, MINDIN |
Location: |
Chr.4:1166932-1208962 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPON2 | NM_001199021.1 | NP_001185950.1 | ||
CB104725.1 | ||||
DA201089.1 | ||||
DB249869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv8891n54 | Chr.4:1181073 - 1261142 on Build GRCh38 | Loss | SPON2 CTBP1-AS CTBP1 LOC101928548 LOC100130872 CTBP1-AS2 |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | TMEM175 RNF212 NKX1-1 PCGF3 MFSD7 CRIPAK LOC100130872 CPLX1 TMED11P SPON2 IDUA DGKQ MAEA PDE6B GAK LOC101928548 FGFRL1 CTBP1-AS2 SLC26A1 CTBP1-AS UVSSA CTBP1 LOC105374344 ATP5I LOC107986211 MYL5 LOC101928521 LOC100129917 |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | NKX1-1 SPON2 CTBP1-AS UVSSA CTBP1 MAEA LOC101928548 CRIPAK LOC100130872 CTBP1-AS2 |
nsv950331 | Chr.4:1164113 - 1239212 on Build GRCh38 | Deletion | SPON2 CTBP1-AS CTBP1 LOC101928548 LOC100130872 |
nsv508988 | Chr.4:1190955 - 1358870 on Build GRCh38 | Insertion | SPON2 CTBP1-AS UVSSA CTBP1 MAEA LOC101928548 LOC100130872 CTBP1-AS2 |
nsv829835 | Chr.4:1103798 - 1310451 on Build GRCh38 | Gain | RNF212 SPON2 CTBP1-AS CTBP1 LOC105374344 MAEA LOC101928548 LOC100130872 TMED11P CTBP1-AS2 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |