Assay Details
Target Gene Details
Entrez Gene ID: | 10861 |
Gene Name: | solute carrier family 26 member 1 |
Gene Aliases: |
CAON, EDM4, SAT-1, SAT1 |
Location: |
Chr.4:979073-993440 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC26A1 | NM_134425.2 | NP_602297.1 | ||
BC015517.2 | AAH15517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv822428 | Chr.4:968896 - 1028144 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1003219 | Chr.4:776106 - 1111676 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3599384 | Chr.4:974637 - 1010875 on Build GRCh38 | Gain |
![]() ![]() |
nsv593226 | Chr.4:948218 - 1025223 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3599385 | Chr.4:983183 - 986434 on Build GRCh38 | Loss |
![]() |
nsv4196 | Chr.4:961755 - 1004599 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469998 | Chr.4:815805 - 1093743 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv1450e212 | Chr.4:976838 - 986842 on Build GRCh38 | Gain |
![]() |
nsv1008352 | Chr.4:819605 - 1041628 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)