Assay Details
Target Gene Details
Entrez Gene ID: | 10144 |
Gene Name: | family with sequence similarity 13 member A |
Gene Aliases: |
ARHGAP48, FAM13A1 |
Location: |
Chr.4:88725954-89111398 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470051 | Chr.4:88723780 - 89721993 on Build GRCh38 | Gain | TIGD2 FAM13A-AS1 FAM13A GPRIN3 |
nsv594806 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain | TIGD2 CCSER1 FAM13A-AS1 MMRN1 FAM13A GPRIN3 HERC3 SNCA-AS1 NAP1L5 SNCA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |