Assay Details
Target Gene Details
Entrez Gene ID: | 93556 |
Gene Name: | EGF like and EMI domain containing 1, pseudogene |
Gene Aliases: |
C3orf50, NCRNA00259 |
Location: |
Chr.3:168249522-168830599 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EGFEM1P | NR_021485.2 | |||
BI461452.1 | ||||
DR000794.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)