Assay Details
Target Gene Details
Entrez Gene ID: | 9940 |
Gene Name: | deleted in lung and esophageal cancer 1 |
Gene Aliases: |
CFAP81, DLC-1, DLC1, F56 |
Location: |
Chr.3:38038595-38122741 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLEC1 | NM_001321153.1 | NP_001308082.1 | ||
NM_007335.3 | NP_031361.2 | |||
NM_007337.3 | NP_031363.2 | |||
XM_006713438.3 | XP_006713501.1 | |||
XM_006713439.3 | XP_006713502.1 | |||
XM_006713440.3 | XP_006713503.1 | |||
XM_011534317.2 | XP_011532619.1 | |||
XM_011534318.2 | XP_011532620.1 | |||
AB020522.1 | BAA77247.1 | |||
AB425198.1 | ||||
BC129824.1 | ||||
BC129825.1 | ||||
BC171799.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv819131 | Chr.3:38037529 - 38044812 on Build GRCh38 | Loss | DLEC1 |
esv2759140 | Chr.3:37789992 - 38062733 on Build GRCh38 | Loss | PLCD1 MIR26A1 DLEC1 ITGA9 CTDSPL VILL ITGA9-AS1 |
nsv590101 | Chr.3:38000691 - 38065526 on Build GRCh38 | Loss | PLCD1 DLEC1 VILL |
nsv428416 | Chr.3:37864474 - 38062733 on Build GRCh38 | Loss | PLCD1 MIR26A1 DLEC1 CTDSPL VILL |
nsv508911 | Chr.3:38019295 - 38091072 on Build GRCh38 | Insertion | PLCD1 DLEC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |