Assay Details
Target Gene Details
Entrez Gene ID: | 254827 |
Gene Name: | N-acetylated alpha-linked acidic dipeptidase-like 2 |
Gene Aliases: |
- |
Location: |
Chr.3:174439669-175809510 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAALADL2 | XM_006713560.2 | XP_006713623.1 | ||
XM_017006075.1 | XP_016861564.1 | |||
XM_017006077.1 | XP_016861566.1 | |||
XM_017006078.1 | XP_016861567.1 | |||
XM_017006080.1 | XP_016861569.1 | |||
XM_017006082.1 | XP_016861571.1 | |||
AJ607398.1 | ||||
AJ607399.1 | ||||
AJ607401.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005730 | Chr.3:174266156 - 174829055 on Build GRCh38 | Gain | NLGN1 NAALADL2 |
nsv829797 | Chr.3:174286339 - 174458042 on Build GRCh38 | Gain | NLGN1 NAALADL2 |
nsv520908 | Chr.3:174083961 - 174816144 on Build GRCh38 | Loss | NLGN1 NAALADL2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |