Assay Details
Target Gene Details
Entrez Gene ID: | 100507086 |
Gene Name: | DLG1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:197298247-197303751 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLG1-AS1 | NR_038289.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3599293 | Chr.3:196759100 - 197444213 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv829830 | Chr.3:197161008 - 197354023 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3893852 | Chr.3:197204950 - 197333165 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3599304 | Chr.3:197291633 - 197300133 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2759214 | Chr.3:197128182 - 197770997 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv818205 | Chr.3:197291254 - 197300056 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv471149 | Chr.3:197183211 - 197621219 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
dgv5038n100 | Chr.3:197134594 - 197659420 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv516873 | Chr.3:197066987 - 197300056 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)