Assay Details
Target Gene Details
Entrez Gene ID: | 5504 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 2 |
Gene Aliases: |
IPP-2, IPP2 |
Location: |
Chr.3:195514422-195543401 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R2 | NM_001291504.1 | NP_001278433.1 | ||
NM_001291505.1 | NP_001278434.1 | |||
NM_001316325.1 | NP_001303254.1 | |||
NM_006241.7 | NP_006232.1 | |||
NR_133014.1 | ||||
XM_006713682.3 | XP_006713745.1 | |||
AJ133812.1 | CAB41680.1 | |||
AY063767.1 | AAL48322.1 | |||
BC007655.2 | AAH07655.1 | |||
BC066921.1 | 2 | 628 | AAH66921.1 | |
BI549862.1 | ||||
BI560631.1 | ||||
X78873.1 | CAA55475.1 | |||
Z29646.1 | CAA82754.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1001387 | Chr.3:195465410 - 195934667 on Build GRCh38 | Gain | PPP1R2 TNK2 LOC101929697 MUC4 LOC105374297 MIR6829 SDHAP2 LOC105374299 MIR570 MUC20 LINC00969 APOD |
nsv1006229 | Chr.3:195350692 - 195747880 on Build GRCh38 | Gain | PPP1R2 MUC4 LOC105374297 SDHAP2 MIR570 MUC20 LINC00969 APOD ACAP2 |
nsv829824 | Chr.3:195410338 - 195559855 on Build GRCh38 | Loss | PPP1R2 ACAP2 |
nsv428430 | Chr.3:195478549 - 196102555 on Build GRCh38 | Gain+Loss | SDHAP1 PPP1R2 TNK2 LOC105374297 MIR6829 SDHAP2 LOC105374299 LOC105374301 MUC20 LINC00969 LOC101929697 MUC4 TFRC MIR570 APOD |
esv2763776 | Chr.3:195514028 - 195775248 on Build GRCh38 | Gain+Loss | PPP1R2 MUC4 LOC105374297 SDHAP2 MIR570 MUC20 LINC00969 APOD |
dgv5014n100 | Chr.3:195514016 - 195747880 on Build GRCh38 | Gain | PPP1R2 MUC4 LOC105374297 SDHAP2 MIR570 MUC20 LINC00969 APOD |
esv2759213 | Chr.3:195382058 - 196344722 on Build GRCh38 | Gain+Loss | SDHAP1 PPP1R2 TNK2 LOC105374297 ZDHHC19 MIR6829 SDHAP2 LOC105374299 TCTEX1D2 LOC105374301 TM4SF19-AS1 MUC20 SLC51A LINC00969 ACAP2 PCYT1A LINC00885 LOC101929697 MUC4 TFRC TM4SF19-TCTEX1D2 MIR570 APOD TM4SF19 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |