Assay Details
Target Gene Details
Entrez Gene ID: | 257313 |
Gene Name: | urotensin 2B |
Gene Aliases: |
U2B, URP, UTS2D |
Location: |
Chr.3:191267155-191330536 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UTS2B | NM_198152.3 | NP_937795.2 | ||
XM_011512631.1 | XP_011510933.1 | |||
XM_017006091.1 | XP_016861580.1 | |||
AB116021.1 | BAC98929.1 | |||
AK090630.1 | ||||
AY321313.1 | AAQ83883.1 | |||
BC112166.1 | AAI12167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428428 | Chr.3:191219045 - 191497807 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2763330 | Chr.3:191265995 - 191273650 on Build GRCh38 | Loss |
![]() |
esv3599095 | Chr.3:191251629 - 191553437 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv2759209 | Chr.3:191149222 - 191561318 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv997716 | Chr.3:191225917 - 191309710 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv3599094 | Chr.3:191225726 - 191310300 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv3599089 | Chr.3:191174866 - 191277287 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv3599099 | Chr.3:191263515 - 191280402 on Build GRCh38 | Gain |
![]() ![]() |
nsv1012488 | Chr.3:191156997 - 191277390 on Build GRCh38 | Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)