Assay Details
Target Gene Details
Entrez Gene ID: | 3568 |
Gene Name: | interleukin 5 receptor subunit alpha |
Gene Aliases: |
CD125, CDw125, HSIL5R3, IL5R |
Location: |
Chr.3:3066324-3110414 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL5RA | NM_000564.4 | NP_000555.2 | ||
NM_001243099.1 | NP_001230028.1 | |||
NM_175726.3 | NP_783853.1 | |||
XM_011533677.2 | XP_011531979.1 | |||
XM_011533678.2 | XP_011531980.1 | |||
AB288089.1 | ||||
AB288090.1 | ||||
AK304256.1 | ||||
M96652.1 | AAA59152.1 | |||
X61176.1 | CAA43483.1 | |||
X61178.1 | CAA43485.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv589371 | Chr.3:2170634 - 3594980 on Build GRCh38 | Loss | CNTN4-AS1 CNTN4 LOC100130207 TRNT1 CRBN IL5RA |
esv2751994 | Chr.3:2200757 - 3178886 on Build GRCh38 | Gain | CNTN4-AS1 CNTN4 TRNT1 CRBN IL5RA |
esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication | CNTN4-AS1 CNTN4 LOC100130207 CNTN4-AS2 TRNT1 CRBN IL5RA |
nsv521796 | Chr.3:3038825 - 3151219 on Build GRCh38 | Loss | CNTN4-AS1 CNTN4 TRNT1 CRBN IL5RA |
dgv4636n100 | Chr.3:2825734 - 3271845 on Build GRCh38 | Gain | CNTN4-AS1 CNTN4 LOC100130207 TRNT1 CRBN IL5RA |
nsv1013843 | Chr.3:3058965 - 3088961 on Build GRCh38 | Loss | CNTN4-AS1 CNTN4 IL5RA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |