Assay Details
Target Gene Details
Entrez Gene ID: | 152137 |
Gene Name: | coiled-coil domain containing 50 |
Gene Aliases: |
C3orf6, DFNA44, YMER |
Location: |
Chr.3:191329082-191398670 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC50 | NM_174908.3 | NP_777568.1 | ||
NM_178335.2 | NP_848018.1 | |||
XM_011512460.1 | XP_011510762.1 | |||
AJ416916.2 | CAC95196.1 | |||
AJ557013.1 | CAD89526.1 | |||
BC065004.1 | AAH65004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv592824 | Chr.3:191343425 - 191355439 on Build GRCh38 | Gain+Loss |
![]() |
nsv428428 | Chr.3:191219045 - 191497807 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
dgv79n17 | Chr.3:191345954 - 191355439 on Build GRCh38 | Loss |
![]() |
esv3599095 | Chr.3:191251629 - 191553437 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3563095 | Chr.3:191346212 - 191354211 on Build GRCh38 | Deletion |
![]() |
esv2759209 | Chr.3:191149222 - 191561318 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
dgv632n27 | Chr.3:191341533 - 191355439 on Build GRCh38 | Loss |
![]() |
dgv8794n54 | Chr.3:191346739 - 191355917 on Build GRCh38 | Loss |
![]() |
dgv114e180 | Chr.3:191346846 - 191353860 on Build GRCh38 | Gain |
![]() |
esv3599104 | Chr.3:191346901 - 191353870 on Build GRCh38 | Loss |
![]() |
dgv132n73 | Chr.3:191346879 - 191354011 on Build GRCh38 | Deletion |
![]() |
nsv10371 | Chr.3:191346459 - 191354045 on Build GRCh38 | Gain+Loss |
![]() |
dgv903e199 | Chr.3:191346896 - 191353886 on Build GRCh38 | Deletion |
![]() |
esv24246 | Chr.3:191346942 - 191353843 on Build GRCh38 | Gain |
![]() |
nsv508266 | Chr.3:191346423 - 191365848 on Build GRCh38 | Deletion |
![]() |
nsv998208 | Chr.3:191303645 - 191457561 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2726395 | Chr.3:191346897 - 191353850 on Build GRCh38 | Deletion |
![]() |
nsv819154 | Chr.3:191346863 - 191353890 on Build GRCh38 | Gain |
![]() |
nsv821209 | Chr.3:191346852 - 191353922 on Build GRCh38 | Deletion |
![]() |
nsv435761 | Chr.3:191346761 - 191356044 on Build GRCh38 | Deletion |
![]() |
nsv965411 | Chr.3:191346443 - 191353939 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)