Assay Details
Target Gene Details
Entrez Gene ID: | 2049 |
Gene Name: | EPH receptor B3 |
Gene Aliases: |
EK2, ETK2, HEK2, TYRO6 |
Location: |
Chr.3:184561799-184582408 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHB3 | NM_004443.3 | NP_004434.2 | ||
BC052968.1 | AAH52968.1 | |||
CB993133.1 | ||||
X75208.1 | CAA53021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3598946 | Chr.3:184450977 - 184897567 on Build GRCh38 | Gain | VPS8 EIF2B5-AS1 MAGEF1 EPHB3 LOC101928992 LOC105374250 |
nsv829814 | Chr.3:184404072 - 184622115 on Build GRCh38 | Loss | EIF2B5-AS1 EPHB3 LOC105374250 |
nsv592671 | Chr.3:184539010 - 184613658 on Build GRCh38 | Loss | EIF2B5-AS1 EPHB3 |
nsv508972 | Chr.3:184538711 - 184616364 on Build GRCh38 | Insertion | EIF2B5-AS1 EPHB3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |