Assay Details
Target Gene Details
Entrez Gene ID: | 4026 |
Gene Name: | LIM domain containing preferred translocation partner in lipoma |
Gene Aliases: |
- |
Location: |
Chr.3:188153050-188890671 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LPP | NM_001167672.2 | NP_001161144.1 | ||
XM_005247446.4 | XP_005247503.1 | |||
XM_005247450.4 | XP_005247507.1 | |||
XM_005247451.4 | XP_005247508.1 | |||
XM_011512820.2 | XP_011511122.1 | |||
XM_011512827.2 | XP_011511129.1 | |||
XM_011512828.2 | XP_011511130.1 | |||
XM_011512831.2 | XP_011511133.1 | |||
XM_011512833.2 | XP_011511135.1 | |||
XM_011512834.2 | XP_011511136.1 | |||
XM_017006377.1 | XP_016861866.1 | |||
XM_017006379.1 | XP_016861868.1 | |||
XM_017006380.1 | XP_016861869.1 | |||
AK304035.1 | ||||
AL833171.1 | ||||
BX474180.1 | ||||
CD697098.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34193 | Chr.3:188129943 - 188550793 on Build GRCh38 | Loss | LPP LPP-AS2 FLJ42393 |
nsv1003486 | Chr.3:188157145 - 188284087 on Build GRCh38 | Gain | LPP FLJ42393 |
esv2751987 | Chr.3:188124933 - 188289510 on Build GRCh38 | Gain | LPP LPP-AS2 FLJ42393 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |