Assay Details
Target Gene Details
Entrez Gene ID: | 152330 |
Gene Name: | contactin 4 |
Gene Aliases: |
AXCAM, BIG-2 |
Location: |
Chr.3:2098771-3059080 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNTN4 | NM_175607.2 | NP_783200.1 | ||
XM_006713004.3 | XP_006713067.1 | |||
XM_011533425.2 | XP_011531727.1 | |||
XM_011533426.2 | XP_011531728.1 | |||
XM_011533427.2 | XP_011531729.1 | |||
XM_017005783.1 | XP_016861272.1 | |||
XM_017005784.1 | XP_016861273.1 | |||
XM_017005785.1 | XP_016861274.1 | |||
AK125460.1 | ||||
AY090737.1 | AAM00025.1 | |||
DA063654.1 | ||||
DA796385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1009942 | Chr.3:788731 - 3038281 on Build GRCh38 | Gain | CNTN4-AS2 CNTN6 CNTN4 LINC01266 |
nsv460321 | Chr.3:1808863 - 2199963 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
esv2751990 | Chr.3:1992731 - 2380637 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
nsv589345 | Chr.3:1708852 - 2712348 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
esv2422409 | Chr.3:1900722 - 2632569 on Build GRCh38 | Deletion | CNTN4-AS2 CNTN4 |
dgv8164n54 | Chr.3:1907610 - 2478272 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
dgv4629n100 | Chr.3:1801319 - 2838526 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
dgv8157n54 | Chr.3:1105517 - 2167148 on Build GRCh38 | Gain | CNTN4-AS2 CNTN6 CNTN4 |
esv3893608 | Chr.3:1662983 - 2177012 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
nsv589363 | Chr.3:1868843 - 2531777 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
esv2422325 | Chr.3:1855887 - 2813527 on Build GRCh38 | Duplication | CNTN4-AS2 CNTN4 |
nsv998345 | Chr.3:1840855 - 2164489 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
nsv460323 | Chr.3:2030247 - 2478272 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication | CNTN4-AS2 TRNT1 LOC100130207 CNTN4 CRBN IL5RA CNTN4-AS1 |
nsv1012867 | Chr.3:2055469 - 2959401 on Build GRCh38 | Loss | CNTN4-AS2 CNTN4 |
nsv1011386 | Chr.3:1987145 - 2359283 on Build GRCh38 | Loss | CNTN4-AS2 CNTN4 |
nsv1006039 | Chr.3:1984601 - 2528799 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
nsv526340 | Chr.3:21733 - 2305176 on Build GRCh38 | Loss | CNTN4-AS2 CNTN6 CHL1-AS1 CHL1 LOC102723448 CNTN4 LOC101927174 LINC01266 |
nsv834600 | Chr.3:1978715 - 2159291 on Build GRCh38 | Loss | CNTN4-AS2 CNTN4 |
nsv460324 | Chr.3:2053858 - 2744818 on Build GRCh38 | Loss | CNTN4-AS2 CNTN4 |
dgv4634n100 | Chr.3:1974634 - 2157221 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
nsv1007298 | Chr.3:1829591 - 2292967 on Build GRCh38 | Loss | CNTN4-AS2 CNTN4 |
nsv589322 | Chr.3:1165012 - 2306437 on Build GRCh38 | Loss | CNTN4-AS2 CNTN6 CNTN4 |
nsv1005807 | Chr.3:1552814 - 2411041 on Build GRCh38 | Gain | CNTN4-AS2 CNTN4 |
esv3575531 | Chr.3:1175929 - 2130831 on Build GRCh38 | Gain | CNTN4-AS2 CNTN6 CNTN4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |