Assay Details
Target Gene Details
Entrez Gene ID: | 7707 |
Gene Name: | zinc finger protein 148 |
Gene Aliases: |
BERF-1, BFCOL1, HT-BETA, ZBP-89, ZFP148, pHZ-52 |
Location: |
Chr.3:125225669-125375354 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF148 | NM_021964.2 | NP_068799.2 | ||
AF039019.1 | AAC39926.1 | |||
AF432210.1 | AAL99917.1 | |||
AJ236885.1 | CAA15422.1 | |||
AK314203.1 | ||||
AL833616.1 | ||||
BC018971.2 | AAH18971.1 | |||
BC035591.1 | AAH35591.1 | |||
BC050260.1 | AAH50260.1 | |||
BQ441085.1 | ||||
DQ090088.1 | AAY99631.1 | |||
L04282.1 | AAA36664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv10311 | Chr.3:125073408 - 125703762 on Build GRCh38 | Gain | OSBPL11 SLC12A8 ZNF148 MIR5092 SNX4 |
nsv1160974 | Chr.3:124577493 - 125300995 on Build GRCh38 | Duplication | KALRN MIR544B MUC13 UMPS SLC12A8 ZNF148 MIR5092 HEG1 ITGB5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |