Assay Details
Target Gene Details
Entrez Gene ID: | 131177 |
Gene Name: | family with sequence similarity 3 member D |
Gene Aliases: |
EF7, OIT1 |
Location: |
Chr.3:58633943-58666843 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM3D | NM_138805.2 | NP_620160.1 | ||
XM_005264862.4 | XP_005264919.1 | |||
XM_005264863.4 | XP_005264920.1 | |||
XM_005264864.4 | XP_005264921.1 | |||
XM_006712965.3 | XP_006713028.1 | |||
XM_006712967.3 | XP_006713030.1 | |||
XM_011533348.2 | XP_011531650.1 | |||
XM_011533349.2 | XP_011531651.1 | |||
XM_011533350.2 | XP_011531652.1 | |||
XM_011533351.2 | XP_011531653.1 | |||
XM_011533352.2 | XP_011531654.1 | |||
XM_011533353.2 | XP_011531655.1 | |||
XM_011533354.2 | XP_011531656.1 | |||
AB105189.1 | ||||
AF494381.1 | AAM94282.1 | |||
AK127546.1 | ||||
AY358677.1 | AAQ89040.1 | |||
BC015359.1 | AAH15359.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4749n100 | Chr.3:58504269 - 60219035 on Build GRCh38 | Gain | LOC339902 FAM3D-AS1 FAM3D FAM107A FHIT C3orf67-AS1 ACOX2 C3orf67 |
nsv460560 | Chr.3:58628289 - 58656725 on Build GRCh38 | Loss | FAM3D-AS1 FAM3D |
esv3596253 | Chr.3:58621679 - 58677821 on Build GRCh38 | Gain | FAM3D-AS1 FAM3D FAM107A |
esv3893702 | Chr.3:58557785 - 58858605 on Build GRCh38 | Loss | FAM3D-AS1 FAM3D FAM107A C3orf67-AS1 C3orf67 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |