Assay Details
Target Gene Details
Entrez Gene ID: | 942 |
Gene Name: | CD86 molecule |
Gene Aliases: |
B7-2, B7.2, B70, CD28LG2, LAB72 |
Location: |
Chr.3:122055362-122121143 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD86 | NM_001206924.1 | NP_001193853.1 | ||
NM_001206925.1 | NP_001193854.1 | |||
NM_175862.4 | NP_787058.4 | |||
AK291497.1 | ||||
AK294663.1 | ||||
AK301237.1 | ||||
AK313103.1 | ||||
AK316203.1 | ||||
BC040261.1 | AAH40261.1 | |||
DA619360.1 | ||||
L25259.1 | AAA58389.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33139 | Chr.3:121781553 - 122372984 on Build GRCh38 | Loss | CD86 IQCB1 SLC15A2 EAF2 CCDC58 CSTA ILDR1 CASR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |