Assay Details
Target Gene Details
Entrez Gene ID: | 10217 |
Gene Name: | CTD small phosphatase like |
Gene Aliases: |
C3orf8, HYA22, PSR1, RBSP3, SCP3 |
Location: |
Chr.3:37862152-37984469 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTDSPL | NM_001008392.1 | NP_001008393.1 | ||
NM_005808.2 | NP_005799.2 | |||
XM_017005519.1 | XP_016861008.1 | |||
XM_017005520.1 | XP_016861009.1 | |||
AI143812.1 | ||||
AI192993.1 | ||||
AJ575644.1 | CAE11804.1 | |||
AJ575645.1 | CAE11805.1 | |||
AY279532.1 | AAP34400.1 | |||
AY364238.1 | AAQ76797.1 | |||
D88153.1 | BAA21667.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759140 | Chr.3:37789992 - 38062733 on Build GRCh38 | Loss | ITGA9-AS1 MIR26A1 ITGA9 CTDSPL VILL DLEC1 PLCD1 |
nsv1160928 | Chr.3:37742341 - 37907632 on Build GRCh38 | Duplication | ITGA9-AS1 ITGA9 CTDSPL |
nsv428416 | Chr.3:37864474 - 38062733 on Build GRCh38 | Loss | MIR26A1 CTDSPL VILL DLEC1 PLCD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |