Assay Details
Target Gene Details
Entrez Gene ID: | 27247 |
Gene Name: | NFU1 iron-sulfur cluster scaffold |
Gene Aliases: |
CGI-33, HIRIP, HIRIP5, MMDS1, NIFUC, Nfu, NifU |
Location: |
Chr.2:69396113-69438123 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFU1 | NM_001002755.2 | NP_001002755.1 | ||
NM_001002756.2 | NP_001002756.1 | |||
NM_015700.3 | NP_056515.2 | |||
NR_045631.1 | ||||
NR_045632.1 | ||||
XM_017003808.1 | XP_016859297.1 | |||
AF132967.1 | AAD27742.1 | |||
AJ132584.1 | CAB53015.1 | |||
AK300700.1 | ||||
AK308567.1 | ||||
AK314004.1 | ||||
AY286306.1 | AAP92372.1 | |||
AY286307.1 | AAP92373.1 | |||
AY335194.1 | AAQ73784.1 | |||
BC113692.1 | ||||
BC113694.1 | ||||
BG385940.1 | ||||
BM846341.1 | ||||
BU178674.1 | ||||
BU507991.1 | ||||
BX538347.1 | CAD98142.1 | |||
DB496651.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33940 | Chr.2:69115288 - 69789041 on Build GRCh38 | Loss | NFU1 AAK1 ANXA4 GFPT1 SNORA36C ANTXR1 |
nsv834252 | Chr.2:69341371 - 69495246 on Build GRCh38 | Loss | NFU1 AAK1 GFPT1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |