Assay Details
Target Gene Details
Entrez Gene ID: | 114818 |
Gene Name: | kelch like family member 29 |
Gene Aliases: |
KBTBD9 |
Location: |
Chr.2:23385419-23708613 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLHL29 | NM_052920.1 | NP_443152.1 | ||
XM_017003264.1 | XP_016858753.1 | |||
BC013982.1 | AAH13982.1 | |||
DR002067.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain | WDCP ATAD2B TP53I3 LOC105374325 FAM228B SF3B6 ITSN2 PFN4 FAM228A MFSD2B KLHL29 UBXN2A FKBP1B |
nsv581173 | Chr.2:23376693 - 23435745 on Build GRCh38 | Loss | KLHL29 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |