Assay Details
Target Gene Details
Entrez Gene ID: | 54842 |
Gene Name: | major facilitator superfamily domain containing 6 |
Gene Aliases: |
MMR2, hMMR2 |
Location: |
Chr.2:190408355-190502315 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MFSD6 | NM_017694.3 | NP_060164.3 | ||
XM_005246654.2 | XP_005246711.1 | |||
XM_005246656.3 | XP_005246713.1 | |||
XM_011511368.1 | XP_011509670.1 | |||
XM_017004392.1 | XP_016859881.1 | |||
AB472299.1 | ||||
AK000167.1 | BAA90987.1 | |||
AK098085.1 | ||||
AK127176.1 | BAC86869.1 | |||
AK225240.1 | ||||
BC050537.2 | AAH50537.2 |
Target Gene Details
Entrez Gene ID: | 100131211 |
Gene Name: | nuclear envelope integral membrane protein 2 |
Gene Aliases: |
TMEM194B |
Location: |
Chr.2:190454088-190607126 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEMP2 | XM_011510458.2 | XP_011508760.1 | ||
XM_017003100.1 | XP_016858589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33134 | Chr.2:190454065 - 190500984 on Build GRCh38 | Gain |
![]() ![]() |
dgv1199e212 | Chr.2:190461225 - 190477989 on Build GRCh38 | Loss |
![]() ![]() |
dgv1198e212 | Chr.2:190461170 - 190469348 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)