Assay Details
Target Gene Details
Entrez Gene ID: | 7535 |
Gene Name: | zeta chain of T cell receptor associated protein kinase 70 |
Gene Aliases: |
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70 |
Location: |
Chr.2:97712030-97744327 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZAP70 | NM_001079.3 | NP_001070.2 | ||
XM_017004867.1 | XP_016860356.1 | |||
XM_017004868.1 | XP_016860357.1 | |||
XM_017004869.1 | XP_016860358.1 | |||
XM_017004870.1 | XP_016860359.1 | |||
AK303341.1 | ||||
AK310628.1 | ||||
AK310637.1 | ||||
BC053878.1 | AAH53878.1 | |||
L05148.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33510 | Chr.2:97700640 - 97724839 on Build GRCh38 | Gain |
![]() ![]() |
nsv2837 | Chr.2:97694463 - 97728803 on Build GRCh38 | Insertion |
![]() ![]() |
esv2759077 | Chr.2:96905372 - 97720044 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)