Assay Details
Target Gene Details
Entrez Gene ID: | 51130 |
Gene Name: | ankyrin repeat and SOCS box containing 3 |
Gene Aliases: |
ASB-3 |
Location: |
Chr.2:53669980-53787009 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASB3 | NM_001201965.1 | NP_001188894.1 | ||
NM_016115.4 | NP_057199.1 | |||
NM_145863.2 | NP_665862.1 | |||
AB209795.1 | BAD93032.1 | |||
AF156778.1 | AAD41895.1 | |||
AK000985.1 | BAA91455.1 | |||
AK001283.1 | BAA91599.1 | |||
AK056069.1 | ||||
AK222595.1 | BAD96315.1 | |||
AK222715.1 | BAD96435.1 | |||
AK303133.1 | ||||
BC006488.2 | AAH06488.1 | |||
BC009569.1 | AAH09569.1 |
Target Gene Details
Entrez Gene ID: | 100302652 |
Gene Name: | GPR75-ASB3 readthrough |
Gene Aliases: |
- |
Location: |
Chr.2:53670293-53860033 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR75-ASB3 | NM_001164165.1 | NP_001157637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3844n100 | Chr.2:53667392 - 53720644 on Build GRCh38 | Loss | ASB3 GPR75-ASB3 |
nsv834058 | Chr.2:53673537 - 53844874 on Build GRCh38 | Gain+Loss | ASB3 ERLEC1 GPR75-ASB3 CHAC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |