Assay Details
Target Gene Details
Entrez Gene ID: | 64135 |
Gene Name: | interferon induced with helicase C domain 1 |
Gene Aliases: |
AGS7, Hlcd, IDDM19, MDA-5, MDA5, RLR-2, SGMRT1 |
Location: |
Chr.2:162267079-162318708 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IFIH1 | NM_022168.3 | NP_071451.2 | ||
AF095844.1 | AAG34368.1 | |||
AK056293.1 | BAB71141.1 | |||
AK292941.1 | ||||
AK314611.1 | ||||
AY017378.1 | AAG54076.1 | |||
BC111750.1 | AAI11751.1 | |||
BQ772836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv583520 | Chr.2:162267541 - 162611723 on Build GRCh38 | Loss | KCNH7 GCA IFIH1 |
dgv749e214 | Chr.2:162204243 - 162287181 on Build GRCh38 | Gain | FAP IFIH1 |
nsv834440 | Chr.2:162145758 - 162318828 on Build GRCh38 | Gain | GCG FAP LOC101929532 GCA IFIH1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |