Assay Details
Target Gene Details
Entrez Gene ID: | 92737 |
Gene Name: | delta/notch like EGF repeat containing |
Gene Aliases: |
UNQ26, bet |
Location: |
Chr.2:229357629-229714570 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNER | NM_139072.3 | NP_620711.3 | ||
XM_005246950.2 | XP_005247007.1 | |||
AF442487.1 | AAM21557.1 | |||
AK289553.1 | ||||
AK304596.1 | ||||
AK304631.1 | ||||
AK308576.1 | ||||
AL137311.1 | CAB70690.1 | |||
AY358891.1 | AAQ89250.1 | |||
BC024766.2 | AAH24766.2 | |||
BC035009.1 | AAH35009.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834564 | Chr.2:229239150 - 229431420 on Build GRCh38 | Gain | DNER PID1 |
nsv3198 | Chr.2:229341777 - 229372836 on Build GRCh38 | Insertion | DNER |
esv3594572 | Chr.2:229347352 - 229417034 on Build GRCh38 | Loss | DNER |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |