Assay Details
Target Gene Details
Entrez Gene ID: | 101928185 |
Gene Name: | uncharacterized LOC101928185 |
Gene Aliases: |
- |
Location: |
Chr.2:132915557-132931494 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928185 | NR_135572.1 | |||
DB058825.1 |
Target Gene Details
Entrez Gene ID: | 344148 |
Gene Name: | NCK associated protein 5 |
Gene Aliases: |
ERIH1, ERIH2, NAP5 |
Location: |
Chr.2:132671788-133675181 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv7087n54 | Chr.2:132915145 - 132916989 on Build GRCh38 | Loss |
![]() ![]() |
esv3592517 | Chr.2:132914339 - 132916963 on Build GRCh38 | Loss |
![]() ![]() |
esv26071 | Chr.2:132914615 - 132917210 on Build GRCh38 | Loss |
![]() ![]() |
nsv583158 | Chr.2:132558519 - 133104527 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv583152 | Chr.2:132369502 - 133631550 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv3592515 | Chr.2:132899712 - 132953048 on Build GRCh38 | Loss |
![]() ![]() |
dgv733e199 | Chr.2:132912041 - 132916906 on Build GRCh38 | Deletion |
![]() ![]() |
esv3560159 | Chr.2:132914254 - 132916929 on Build GRCh38 | Deletion |
![]() ![]() |
esv3592516 | Chr.2:132912099 - 132916928 on Build GRCh38 | Loss |
![]() ![]() |
esv2670272 | Chr.2:132914292 - 132916871 on Build GRCh38 | Deletion |
![]() ![]() |
nsv1004604 | Chr.2:132900543 - 133004221 on Build GRCh38 | Loss |
![]() ![]() |
nsv583159 | Chr.2:132909712 - 132920884 on Build GRCh38 | Loss |
![]() ![]() |
nsv1011421 | Chr.2:132137668 - 132924612 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)