Assay Details
Target Gene Details
Entrez Gene ID: | 130507 |
Gene Name: | ubiquitin protein ligase E3 component n-recognin 3 (putative) |
Gene Aliases: |
ZNF650 |
Location: |
Chr.2:169827463-170084131 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBR3 | NM_172070.3 | NP_742067.3 | ||
XM_005246299.4 | XP_005246356.1 | |||
XM_005246300.4 | XP_005246357.1 | |||
XM_006712268.3 | XP_006712331.1 | |||
XM_006712269.3 | XP_006712332.1 | |||
XM_006712270.3 | XP_006712333.2 | |||
XM_006712271.3 | XP_006712334.1 | |||
XM_011510604.2 | XP_011508906.1 | |||
XM_017003356.1 | XP_016858845.1 | |||
XM_017003357.1 | XP_016858846.1 | |||
XM_017003358.1 | XP_016858847.1 | |||
XM_017003359.1 | XP_016858848.1 | |||
BF446017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv583608 | Chr.2:169769815 - 169984892 on Build GRCh38 | Gain | UBR3 METTL5 SSB CYB5AP2 |
nsv1006657 | Chr.2:169863723 - 170244999 on Build GRCh38 | Gain | UBR3 MYO3B |
nsv10201 | Chr.2:169701593 - 170169904 on Build GRCh38 | Loss | KLHL23 UBR3 METTL5 PHOSPHO2-KLHL23 SSB CYB5AP2 PHOSPHO2 |
nsv1011368 | Chr.2:169423740 - 170042296 on Build GRCh38 | Loss | KLHL23 UBR3 METTL5 CCDC173 PHOSPHO2-KLHL23 FASTKD1 KLHL41 SSB CYB5AP2 PPIG PHOSPHO2 BBS5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |