Assay Details
Target Gene Details
Entrez Gene ID: | 647012 |
Gene Name: | YY1 transcription factor pseudogene 2 |
Gene Aliases: |
- |
Location: |
Chr.2:138897324-138899174 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YY1P2 | NR_033658.1 | 1 | 306 | |
BI915156.1 | 1 | 306 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv583183 | Chr.2:138887302 - 138956334 on Build GRCh38 | Loss | YY1P2 |
esv3592648 | Chr.2:138894525 - 138900390 on Build GRCh38 | Loss | YY1P2 |