Assay Details
Target Gene Details
Entrez Gene ID: | 2641 |
Gene Name: | glucagon |
Gene Aliases: |
GLP1, GLP2, GRPP |
Location: |
Chr.2:162142869-162152404 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GCG | NM_002054.4 | NP_002045.1 | ||
BC005278.1 | AAH05278.1 | |||
BM272442.1 | ||||
J04040.1 | AAA52567.1 |
Target Gene Details
Entrez Gene ID: | 101929532 |
Gene Name: | uncharacterized LOC101929532 |
Gene Aliases: |
- |
Location: |
Chr.2:162114441-162172739 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929532 | NR_110255.1 | |||
BX648358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834440 | Chr.2:162145758 - 162318828 on Build GRCh38 | Gain | GCA LOC101929532 IFIH1 FAP GCG |
nsv834439 | Chr.2:162057118 - 162223456 on Build GRCh38 | Gain | LOC101929532 FAP DPP4 GCG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |