Assay Details
Target Gene Details
Entrez Gene ID: | 105373944 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.2:236161340-236164886 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105373944 | XM_017005449.1 | XP_016860938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv821922 | Chr.2:236107343 - 236246934 on Build GRCh38 | Gain | LOC105373944 GBX2 AGAP1 ASB18 |
nsv821924 | Chr.2:236162662 - 236172520 on Build GRCh38 | Gain | LOC105373944 GBX2 |
nsv821923 | Chr.2:236134173 - 236228146 on Build GRCh38 | Gain | LOC105373944 GBX2 ASB18 |
nsv953211 | Chr.2:236158057 - 236174757 on Build GRCh38 | Deletion | LOC105373944 GBX2 |
nsv525913 | Chr.2:236153310 - 236174434 on Build GRCh38 | Loss | LOC105373944 GBX2 |
nsv526683 | Chr.2:236143831 - 236165133 on Build GRCh38 | Loss | LOC105373944 |
nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss | RAMP1 LOC105373944 GBX2 LRRFIP1 LOC643387 ESPNL KLHL30 MLPH TRAF3IP1 RBM44 LOC105373991 LOC93463 IQCA1 AGAP1 RAB17 HES6 LINC01107 ERFE LOC101927958 LOC105373958 COL6A3 SCLY UBE2F-SCLY ASB1 ACKR3 ILKAP PER2 MIR6811 COPS8 UBE2F PRLH LOC151174 ASB18 |
esv2751845 | Chr.2:235963461 - 236174434 on Build GRCh38 | Gain | LOC105373944 GBX2 AGAP1 |
esv33938 | Chr.2:236043820 - 236294716 on Build GRCh38 | Loss | LOC105373944 GBX2 AGAP1 ASB18 |