Assay Details
Target Gene Details
Entrez Gene ID: | 150946 |
Gene Name: | GRB2 associated regulator of MAPK1 subtype 2 |
Gene Aliases: |
FAM59B, GAREML |
Location: |
Chr.2:26173091-26202405 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GAREM2 | NM_001168241.1 | NP_001161713.1 | ||
NM_001191033.1 | NP_001177962.1 | |||
XM_006711951.3 | XP_006712014.1 | |||
XM_011532564.2 | XP_011530866.1 | |||
XM_011532565.2 | XP_011530867.1 | |||
XM_011532566.2 | XP_011530868.1 | |||
XM_011532567.2 | XP_011530869.1 | |||
AB124552.1 | ||||
AK090454.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2648 | Chr.2:26177840 - 26222776 on Build GRCh38 | Deletion | GAREM2 HADHA |
nsv520638 | Chr.2:26169872 - 26190499 on Build GRCh38 | Loss | GAREM2 |
esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss | DTNB GAREM2 ADGRF3 EPT1 HADHB ASXL2 OTOF DRC1 C2orf70 CIB4 HADHA KIF3C RAB10 |