Assay Details
Target Gene Details
Entrez Gene ID: | 129563 |
Gene Name: | DIS3 like 3'-5' exoribonuclease 2 |
Gene Aliases: |
FAM6A, PRLMNS, hDIS3L2 |
Location: |
Chr.2:231961583-232343968 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DIS3L2 | NM_001257281.1 | NP_001244210.1 | ||
NM_001257282.1 | NP_001244211.1 | |||
NM_152383.4 | NP_689596.4 | |||
NR_046476.1 | ||||
NR_046477.1 | ||||
AF443854.1 | AAP97321.1 | |||
AK094293.1 | BAC04324.1 | |||
AK295363.1 | ||||
BC026166.1 | AAH26166.1 | |||
BC036113.1 | AAH36113.2 | |||
BX648325.1 | CAH10545.1 | |||
DA058505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759117 | Chr.2:231672114 - 232141148 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)