Assay Details
Target Gene Details
Entrez Gene ID: | 65220 |
Gene Name: | NAD kinase |
Gene Aliases: |
dJ283E3.1 |
Location: |
Chr.1:1751232-1780514 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NADK | NM_001198993.1 | NP_001185922.1 | ||
NM_001198994.1 | NP_001185923.1 | |||
NM_023018.4 | NP_075394.3 | |||
XM_005244778.2 | XP_005244835.1 | |||
XM_006710837.2 | XP_006710900.1 | |||
XM_006710838.2 | XP_006710901.1 | |||
XM_006710839.1 | XP_006710902.1 | |||
XM_011542006.1 | XP_011540308.1 | |||
XM_011542007.1 | XP_011540309.1 | |||
XM_011542008.1 | XP_011540310.1 | |||
XM_011542009.2 | XP_011540311.1 | |||
XM_017002143.1 | XP_016857632.1 | |||
XM_017002145.1 | XP_016857634.1 | |||
XM_017002146.1 | XP_016857635.1 | |||
AF250320.1 | AAG44568.1 | |||
AK023114.1 | BAB14412.1 | |||
AK290161.1 | ||||
AK291599.1 | ||||
AK291728.1 | ||||
AK307645.1 | ||||
AY090771.1 | AAM01195.1 | |||
BC001709.1 | AAH01709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003545 | Chr.1:1696467 - 1760882 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
dgv5n111 | Chr.1:1575421 - 1823403 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834091 | Chr.1:1701359 - 1880638 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv545041 | Chr.1:1753033 - 1780259 on Build GRCh38 | Loss |
![]() |
nsv545030 | Chr.1:1689422 - 1774697 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv10161 | Chr.1:776731 - 1777210 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160797 | Chr.1:1566216 - 1805918 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2758913 | Chr.1:1389149 - 2092899 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)