Assay Details
Target Gene Details
Entrez Gene ID: | 65220 |
Gene Name: | NAD kinase |
Gene Aliases: |
dJ283E3.1 |
Location: |
Chr.1:1751232-1780514 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NADK | NM_001198993.1 | NP_001185922.1 | ||
NM_001198994.1 | NP_001185923.1 | |||
NM_023018.4 | NP_075394.3 | |||
XM_005244778.2 | XP_005244835.1 | |||
XM_006710837.2 | XP_006710900.1 | |||
XM_006710838.2 | XP_006710901.1 | |||
XM_006710839.1 | XP_006710902.1 | |||
XM_011542006.1 | XP_011540308.1 | |||
XM_011542007.1 | XP_011540309.1 | |||
XM_011542008.1 | XP_011540310.1 | |||
XM_011542009.2 | XP_011540311.1 | |||
XM_017002143.1 | XP_016857632.1 | |||
XM_017002145.1 | XP_016857634.1 | |||
XM_017002146.1 | XP_016857635.1 | |||
AF250320.1 | AAG44568.1 | |||
AK023114.1 | BAB14412.1 | |||
AK290161.1 | ||||
AK291599.1 | ||||
AK291728.1 | ||||
BC001709.1 | AAH01709.1 | |||
CN349189.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5n111 | Chr.1:1575421 - 1823403 on Build GRCh38 | Deletion | NADK CDK11A MMP23A MMP23B SLC35E2B GNB1 SLC35E2 CDK11B FNDC10 MIB2 |
nsv834091 | Chr.1:1701359 - 1880638 on Build GRCh38 | Loss | NADK CDK11A MMP23A GNB1 SLC35E2 |
nsv545041 | Chr.1:1753033 - 1780259 on Build GRCh38 | Loss | NADK |
nsv545030 | Chr.1:1689422 - 1774697 on Build GRCh38 | Gain | NADK CDK11A MMP23A SLC35E2B SLC35E2 |
nsv10161 | Chr.1:776731 - 1777210 on Build GRCh38 | Gain+Loss | AGRN RNF223 MXRA8 SAMD11 MRPL20 FAM87B CPSF3L FNDC10 MIB2 HES4 ACAP3 NOC2L LOC107985729 AURKAIP1 B3GALT6 TNFRSF18 TMEM88B MIR200B LINC01342 MIR6808 DVL1 PUSL1 LOC100130417 LOC100288069 MIR6727 ANKRD65 LINC01128 LOC148413 MMP23B LOC107985728 SLC35E2B KLHL17 LOC284600 TTLL10 LOC100287934 NADK CDK11A SSU72 UBE2J2 CDK11B ATAD3A TTLL10-AS1 PERM1 TMEM240 ATAD3B VWA1 MIR6726 LOC102724312 SLC35E2 LINC00115 FAM41C MMP23A TNFRSF4 CPTP LOC105378948 FAM132A MIR429 CCNL2 TAS1R3 LOC100288175 PLEKHN1 MIR200A C1orf159 SDF4 ATAD3C ISG15 SCNN1D |
nsv1160797 | Chr.1:1566216 - 1805918 on Build GRCh38 | Duplication | NADK CDK11A MMP23A MMP23B SLC35E2B GNB1 SSU72 SLC35E2 CDK11B FNDC10 MIB2 |
nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss | DDX11L1 RNF223 MXRA8 SAMD11 OR4F16 CPSF3L ACAP3 MTND1P23 LOC107985729 LOC729737 TNFRSF18 MIR200B MIR6859-2 LOC105378589 DVL1 CFAP74 MIR6727 OR4F29 LOC107985728 SLC35E2B TMEM52 MIR6723 LOC100287934 CDK11A SSU72 CALML6 ATAD3B VWA1 LOC102724312 SLC35E2 LOC101928626 LINC00115 FAM41C LOC107984841 GABRD LOC105378592 TAS1R3 MIR1302-2 LOC100288175 PLEKHN1 MIR6859-1 SKI MORN1 SDF4 ATAD3C SCNN1D AGRN LOC100506504 MRPL20 PRKCZ LOC105378949 FAM87B FNDC10 MIB2 HES4 NOC2L AURKAIP1 B3GALT6 FAM138A TMEM88B LOC102725121 MTND2P28 WASH7P LINC01342 MIR6808 PUSL1 LOC100130417 LOC100288069 ANKRD65 LINC01128 LOC148413 MMP23B KLHL17 GNB1 LOC284600 TTLL10 NADK LOC100129534 UBE2J2 LOC100133331 OR4F5 CDK11B ATAD3A TTLL10-AS1 PERM1 TMEM240 MIR6726 MMP23A TNFRSF4 FAAP20 CPTP LOC105378948 LOC105378591 FAM132A MIR429 CCNL2 LOC105378947 MIR200A C1orf159 LOC100134822 LOC100132287 ISG15 |
esv2758913 | Chr.1:1389149 - 2092899 on Build GRCh38 | Gain+Loss | NADK CDK11A MRPL20 PRKCZ SSU72 LOC105378949 CALML6 CDK11B ATAD3A FNDC10 MIB2 TMEM240 ATAD3B VWA1 LOC107985729 LOC102724312 SLC35E2 TMEM88B MMP23A LOC105378589 LOC105378591 GABRD CCNL2 CFAP74 ANKRD65 LOC148413 MMP23B SLC35E2B GNB1 ATAD3C TMEM52 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |