Assay Details
Target Gene Details
Entrez Gene ID: | 148581 |
Gene Name: | ubiquitin conjugating enzyme E2 U (putative) |
Gene Aliases: |
- |
Location: |
Chr.1:64202802-64267381 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE2U | NM_152489.1 | NP_689702.1 | ||
XM_005270498.4 | XP_005270555.1 | |||
XM_006710379.3 | XP_006710442.1 | |||
XM_011540764.2 | XP_011539066.1 | |||
XM_017000379.1 | XP_016855868.1 | |||
XM_017000380.1 | XP_016855869.1 | |||
XM_017000381.1 | XP_016855870.1 | |||
XM_017000382.1 | XP_016855871.1 | |||
XM_017000383.1 | XP_016855872.1 | |||
XM_017000384.1 | XP_016855873.1 | |||
XM_017000385.1 | XP_016855874.1 | |||
BC029895.1 | AAH29895.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546436 | Chr.1:64205389 - 64269305 on Build GRCh38 | Gain |
![]() |
esv2758938 | Chr.1:64211465 - 64310713 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)