Assay Details
Target Gene Details
Entrez Gene ID: | 58512 |
Gene Name: | DLG associated protein 3 |
Gene Aliases: |
DAP3, SAPAP3 |
Location: |
Chr.1:34862120-34929585 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP3 | NM_001080418.2 | NP_001073887.1 | ||
XM_011541879.2 | XP_011540181.1 | |||
XM_011541880.2 | XP_011540182.1 | |||
AF131778.1 | AAD20042.1 | |||
BC114370.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012995 | Chr.1:34786068 - 34884653 on Build GRCh38 | Gain | DLGAP3 GJB3 SMIM12 GJA4 |
esv3577027 | Chr.1:34867607 - 34874000 on Build GRCh38 | Gain | DLGAP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |