Assay Details
Target Gene Details
Entrez Gene ID: | 7818 |
Gene Name: | death associated protein 3 |
Gene Aliases: |
DAP-3, MRP-S29, MRPS29, bMRP-10 |
Location: |
Chr.1:155688104-155739010 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3578199 | Chr.1:155687166 - 155693640 on Build GRCh38 | Loss |
![]() ![]() |
nsv436522 | Chr.1:155589722 - 155704458 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() |
esv3587631 | Chr.1:155689908 - 155694673 on Build GRCh38 | Loss |
![]() |
nsv514012 | Chr.1:155689985 - 155694417 on Build GRCh38 | Loss |
![]() |
esv28903 | Chr.1:155689963 - 155694691 on Build GRCh38 | Loss |
![]() |
esv3587628 | Chr.1:155619701 - 155695480 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv428246 | Chr.1:155588406 - 155816040 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv33869 | Chr.1:155253492 - 155948170 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2718429 | Chr.1:155648347 - 155763436 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2662699 | Chr.1:155689968 - 155694673 on Build GRCh38 | Deletion |
![]() |
nsv3054 | Chr.1:155684044 - 155703718 on Build GRCh38 | Deletion |
![]() ![]() |
nsv547965 | Chr.1:155593361 - 155718809 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)