Assay Details
Target Gene Details
Entrez Gene ID: | 7161 |
Gene Name: | tumor protein p73 |
Gene Aliases: |
P73 |
Location: |
Chr.1:3652565-3736201 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TP73 | NM_001204184.1 | NP_001191113.1 | ||
NM_001204185.1 | NP_001191114.1 | |||
NM_001204186.1 | NP_001191115.1 | |||
NM_001204187.1 | NP_001191116.1 | |||
NM_001204188.1 | NP_001191117.1 | |||
NM_005427.3 | NP_005418.1 | |||
BC117251.1 | ||||
BC117253.1 | ||||
Y11416.1 | CAA72220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470687 | Chr.1:3356499 - 3763210 on Build GRCh38 | Loss | MEGF6 WRAP73 MIR551A ARHGEF16 CCDC27 TPRG1L TP73 PRDM16 TP73-AS1 |
nsv545128 | Chr.1:3630371 - 3700037 on Build GRCh38 | Loss | WRAP73 TP73 |
nsv1001715 | Chr.1:3627619 - 3769754 on Build GRCh38 | Loss | WRAP73 CCDC27 TPRG1L TP73 TP73-AS1 |
esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain | LINC01134 SMIM1 DFFB TPRG1L TP73-AS1 LRRC47 MEGF6 WRAP73 C1orf174 CCDC27 LINC01346 TP73 CEP104 |
nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain | LINC01134 SMIM1 AJAP1 DFFB TPRG1L TP73-AS1 LRRC47 MEGF6 WRAP73 MIR551A ARHGEF16 LOC284661 C1orf174 CCDC27 LINC01346 TP73 CEP104 |
nsv951959 | Chr.1:3051037 - 3812036 on Build GRCh38 | Deletion | LINC00982 SMIM1 TPRG1L TP73-AS1 LRRC47 MEGF6 WRAP73 MIR551A LOC105378604 ARHGEF16 CCDC27 MIR4251 TP73 PRDM16 |
nsv830314 | Chr.1:3660776 - 3804275 on Build GRCh38 | Gain+Loss | LRRC47 SMIM1 CCDC27 TP73 TP73-AS1 |
esv3585045 | Chr.1:3611161 - 3686920 on Build GRCh38 | Gain | MEGF6 WRAP73 TPRG1L TP73 |
esv2758916 | Chr.1:3668477 - 3738972 on Build GRCh38 | Gain | TP73 TP73-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |