Assay Details
Target Gene Details
Entrez Gene ID: | 280 |
Gene Name: | amylase, alpha 2B (pancreatic) |
Gene Aliases: |
AMY2, AMY3, HXA |
Location: |
Chr.1:103554644-103579534 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMY2B | NM_020978.4 | NP_066188.1 | ||
AK095605.1 | ||||
BC011179.1 | AAH11179.1 | |||
BX456665.2 | ||||
DA157436.1 | ||||
DB235608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526367 | Chr.1:103477009 - 103787255 on Build GRCh38 | Gain | AMY2B RNPC3 AMY1C LOC101928436 ACTG1P4 AMY1A AMY2A AMY1B |
esv3893856 | Chr.1:103555093 - 103755245 on Build GRCh38 | Gain+Loss | AMY2B RNPC3 AMY1C ACTG1P4 AMY1A AMY2A AMY1B |
dgv35e214 | Chr.1:103560121 - 103594054 on Build GRCh38 | Gain | AMY2B ACTG1P4 |
dgv218n100 | Chr.1:103556018 - 103725588 on Build GRCh38 | Gain | AMY2B ACTG1P4 AMY1A AMY2A AMY1B |
dgv213n100 | Chr.1:103458409 - 103725588 on Build GRCh38 | Gain | AMY2B RNPC3 LOC101928436 ACTG1P4 AMY1A AMY2A AMY1B |
nsv830904 | Chr.1:103393009 - 103564563 on Build GRCh38 | Loss | AMY2B RNPC3 LOC101928436 |
dgv14n21 | Chr.1:103374133 - 103586151 on Build GRCh38 | Gain | AMY2B RNPC3 LOC101928436 ACTG1P4 |
nsv428157 | Chr.1:103447737 - 103909380 on Build GRCh38 | Gain+Loss | AMY2B RNPC3 AMY1C LOC101928436 ACTG1P4 AMY1A AMY2A AMY1B |
nsv830915 | Chr.1:103482208 - 103750660 on Build GRCh38 | Gain+Loss | AMY2B RNPC3 AMY1C LOC101928436 ACTG1P4 AMY1A AMY2A AMY1B |
dgv2e203 | Chr.1:103556030 - 103761017 on Build GRCh38 | Gain+Loss | AMY2B AMY1C ACTG1P4 AMY1A AMY2A AMY1B |
esv3583026 | Chr.1:103524036 - 103589563 on Build GRCh38 | Gain | AMY2B RNPC3 LOC101928436 ACTG1P4 |
nsv523177 | Chr.1:103500563 - 103586151 on Build GRCh38 | Gain | AMY2B RNPC3 LOC101928436 ACTG1P4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |