Assay Details
Target Gene Details
Entrez Gene ID: | 375033 |
Gene Name: | platelet endothelial aggregation receptor 1 |
Gene Aliases: |
JEDI, MEGF12 |
Location: |
Chr.1:156893697-156916434 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PEAR1 | NM_001080471.1 | NP_001073940.1 | ||
XM_005245141.3 | XP_005245198.1 | |||
XM_011509510.2 | XP_011507812.1 | |||
XM_011509511.2 | XP_011507813.1 | |||
XM_011509512.2 | XP_011507814.1 | |||
XM_017001236.1 | XP_016856725.1 | |||
XM_017001237.1 | XP_016856726.1 | |||
XM_017001238.1 | XP_016856727.1 | |||
XM_017001239.1 | XP_016856728.1 | |||
XM_017001242.1 | XP_016856731.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831648 | Chr.1:156694085 - 156912126 on Build GRCh38 | Loss | ISG20L2 RRNAD1 CRABP2 INSRR SH2D2A PEAR1 HDGF PRCC NTRK1 MRPL24 |
nsv831659 | Chr.1:156771159 - 156980204 on Build GRCh38 | Loss | MIR765 INSRR SH2D2A LRRC71 PEAR1 PRCC NTRK1 ARHGEF11 |
nsv1006558 | Chr.1:156877325 - 156907288 on Build GRCh38 | Gain | PEAR1 NTRK1 |
nsv998265 | Chr.1:156834329 - 156903671 on Build GRCh38 | Gain | INSRR PEAR1 NTRK1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |