Assay Details
Target Gene Details
Entrez Gene ID: | 84838 |
Gene Name: | zinc finger protein 496 |
Gene Aliases: |
NIZP1, ZFP496, ZKSCAN17, ZSCAN49 |
Location: |
Chr.1:247297413-247333376 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833137 | Chr.1:247274015 - 247444126 on Build GRCh38 | Gain | LOC107985115 NLRP3 ZNF496 |
nsv833126 | Chr.1:247109782 - 247312848 on Build GRCh38 | Loss | VN1R5 ZNF124 MIR3916 ZNF496 C1orf229 |
esv33129 | Chr.1:246911128 - 248165533 on Build GRCh38 | Gain+Loss | GCSAML OR1C1 OR2T8 LOC107985115 NLRP3 OR6F1 TRIM58 OR2G3 OR2AK2 ZNF124 ZNF670-ZNF695 ZNF669 OR9H1P AHCTF1 MIR3916 OR2M5 OR2L2 OR2B11 ZNF496 OR2L8 OR2L13 OR2G2 OR2W5 OR2M1P VN1R5 GCSAML-AS1 OR11L1 ZNF670 OR13G1 OR2L3 OR2L5 OR2L1P OR2W3 C1orf229 OR14A16 ZNF695 OR2C3 |
esv3589514 | Chr.1:247297392 - 247354590 on Build GRCh38 | Gain | LOC107985115 ZNF496 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |