Assay Details
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4n111 | Chr.1:869076 - 914637 on Build GRCh38 | Duplication | LOC284600 FAM41C |
nsv1004962 | Chr.1:629241 - 915295 on Build GRCh38 | Gain | LOC107984841 LOC100133331 OR4F16 LINC00115 LOC284600 MIR6723 LOC100287934 FAM41C MTND2P28 LINC01128 MTND1P23 LOC100288069 FAM87B |
nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss | MIR429 LOC100134822 LOC105378947 OR4F5 MIR200A MORN1 PLEKHN1 TMEM52 LOC105378949 MXRA8 MTND2P28 SDF4 MIR200B DDX11L1 KLHL17 MMP23B LOC100130417 MIR6726 FAAP20 ATAD3B SLC35E2 VWA1 LOC105378589 OR4F29 DVL1 MIR6727 ISG15 AURKAIP1 CPTP MTND1P23 TTLL10 LOC100133331 WASH7P TMEM88B LOC101928626 CDK11A SKI FAM41C PRKCZ LOC102724312 LINC01342 GNB1 TTLL10-AS1 MIR6859-1 SSU72 RNF223 PUSL1 LOC100288069 FAM87B CPSF3L LOC100288175 MMP23A LINC00115 LOC729737 LOC284600 LOC100506504 LOC102725121 B3GALT6 TNFRSF4 TNFRSF18 SCNN1D MIB2 FAM132A CCNL2 FNDC10 PERM1 HES4 SLC35E2B UBE2J2 NOC2L LOC107984841 OR4F16 LOC107985729 LOC105378592 SAMD11 LOC100132287 LOC107985728 CALML6 MIR6723 CFAP74 MIR6808 FAM138A ACAP3 TAS1R3 LOC148413 CDK11B LOC100287934 C1orf159 MIR1302-2 ATAD3A LINC01128 TMEM240 LOC105378948 ANKRD65 AGRN GABRD NADK LOC100129534 LOC105378591 MIR6859-2 ATAD3C MRPL20 |
nsv950451 | Chr.1:899421 - 1094520 on Build GRCh38 | Deletion | LOC100288175 KLHL17 LOC284600 LOC105378948 AGRN SAMD11 LOC100130417 LOC107985728 PERM1 PLEKHN1 HES4 C1orf159 RNF223 ISG15 NOC2L |
nsv509035 | Chr.1:891406 - 985724 on Build GRCh38 | Insertion | PERM1 PLEKHN1 KLHL17 LOC284600 SAMD11 LOC100130417 NOC2L LOC107985728 |
dgv1n67 | Chr.1:811028 - 914557 on Build GRCh38 | Gain | LINC00115 LOC284600 FAM41C LINC01128 FAM87B |
nsv1005935 | Chr.1:805477 - 915295 on Build GRCh38 | Gain | LINC00115 LOC284600 FAM41C LINC01128 FAM87B |
esv3584982 | Chr.1:837710 - 917284 on Build GRCh38 | Gain | LOC284600 FAM41C LINC01128 LOC100130417 |
nsv1009537 | Chr.1:841742 - 915295 on Build GRCh38 | Gain | LOC284600 FAM41C LINC01128 |
nsv428334 | Chr.1:874371 - 1220569 on Build GRCh38 | Gain | LOC100288175 MIR429 LOC284600 MIR200A TNFRSF4 TNFRSF18 PERM1 PLEKHN1 HES4 C1orf159 FAM41C SDF4 MIR200B NOC2L KLHL17 LOC105378948 AGRN SAMD11 LOC100130417 LOC107985728 LINC01342 TTLL10-AS1 RNF223 ISG15 TTLL10 |
nsv10161 | Chr.1:776731 - 1777210 on Build GRCh38 | Gain+Loss | LOC100288175 MIR429 MMP23A LINC00115 LOC284600 B3GALT6 MIR200A TNFRSF4 TNFRSF18 SCNN1D MIB2 FAM132A CCNL2 FNDC10 PERM1 PLEKHN1 HES4 SLC35E2B UBE2J2 MXRA8 SDF4 MIR200B NOC2L KLHL17 MMP23B LOC107985729 SAMD11 LOC100130417 LOC107985728 MIR6726 ATAD3B SLC35E2 VWA1 DVL1 MIR6727 ISG15 AURKAIP1 CPTP TTLL10 MIR6808 ACAP3 TMEM88B TAS1R3 LOC148413 CDK11A CDK11B LOC100287934 C1orf159 FAM41C ATAD3A LINC01128 TMEM240 LOC105378948 ANKRD65 AGRN NADK LOC102724312 LINC01342 TTLL10-AS1 ATAD3C MRPL20 SSU72 RNF223 PUSL1 LOC100288069 FAM87B CPSF3L |
dgv5n100 | Chr.1:585989 - 1114424 on Build GRCh38 | Gain | LOC100288175 LOC100133331 LINC00115 LOC101928626 LOC284600 LOC105378947 PERM1 PLEKHN1 HES4 LOC100287934 C1orf159 FAM41C MTND2P28 LINC01128 NOC2L LOC107984841 OR4F16 KLHL17 LOC105378948 AGRN SAMD11 LOC100130417 LOC107985728 MIR6723 RNF223 ISG15 MTND1P23 LOC100288069 FAM87B |
nsv517709 | Chr.1:817186 - 1275912 on Build GRCh38 | Gain+Loss | LOC100288175 MIR429 LINC00115 LOC284600 B3GALT6 MIR200A TNFRSF4 TNFRSF18 FAM132A PERM1 PLEKHN1 HES4 C1orf159 UBE2J2 FAM41C SDF4 LINC01128 MIR200B NOC2L KLHL17 LOC105378948 AGRN SAMD11 LOC100130417 LOC107985728 LINC01342 TTLL10-AS1 RNF223 ISG15 TTLL10 FAM87B |
dgv1n111 | Chr.1:690090 - 939522 on Build GRCh38 | Duplication | LOC107984841 LOC100133331 OR4F16 LINC00115 LOC284600 SAMD11 LOC100130417 LOC107985728 LOC100287934 FAM41C LINC01128 LOC100288069 FAM87B |
dgv3n111 | Chr.1:783283 - 939522 on Build GRCh38 | Duplication | LINC00115 LOC284600 LOC100287934 SAMD11 FAM41C LINC01128 LOC100130417 LOC107985728 FAM87B |
dgv2n67 | Chr.1:877618 - 1426500 on Build GRCh38 | Gain | LOC100288175 MIR429 ACAP3 TMEM88B LOC284600 TAS1R3 LOC148413 B3GALT6 MIR200A TNFRSF4 TNFRSF18 SCNN1D FAM132A CCNL2 PERM1 PLEKHN1 HES4 C1orf159 UBE2J2 MXRA8 SDF4 MIR200B NOC2L KLHL17 LOC105378948 ANKRD65 AGRN SAMD11 LOC100130417 LOC107985728 MIR6726 LINC01342 TTLL10-AS1 MRPL20 DVL1 RNF223 MIR6727 ISG15 AURKAIP1 PUSL1 CPTP TTLL10 MIR6808 CPSF3L |
nsv832980 | Chr.1:848279 - 1007037 on Build GRCh38 | Gain | PERM1 PLEKHN1 KLHL17 LOC284600 HES4 SAMD11 FAM41C LINC01128 LOC100130417 NOC2L LOC107985728 |
esv3584976 | Chr.1:733250 - 914824 on Build GRCh38 | Gain | LOC107984841 LINC00115 LOC284600 LOC100287934 FAM41C LINC01128 LOC100288069 FAM87B |