Assay Details
Target Gene Details
Entrez Gene ID: | 7159 |
Gene Name: | tumor protein p53 binding protein 2 |
Gene Aliases: |
53BP2, ASPP2, BBP, P53BP2, PPP1R13A |
Location: |
Chr.1:223779893-223845981 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TP53BP2 | NM_001031685.2 | NP_001026855.2 | ||
NM_005426.2 | NP_005417.1 | |||
XM_011544268.1 | XP_011542570.1 | |||
XM_011544269.2 | XP_011542571.1 | |||
XM_017002220.1 | XP_016857709.1 | |||
AJ318888.2 | CAC83012.1 | |||
AK294432.1 | ||||
AK294654.1 | ||||
AK294964.1 | ||||
AK295378.1 | ||||
AK316016.1 | ||||
AK316103.1 | ||||
AL357233.1 | ||||
BC040247.1 | ||||
BC058918.1 | ||||
U09582.1 | AAA21597.1 | |||
U58334.1 | AAC50557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34082 | Chr.1:223663316 - 223956807 on Build GRCh38 | Loss | CAPN8 TP53BP2 CAPN2 GTF2IP20 |
esv2759000 | Chr.1:223747518 - 224175461 on Build GRCh38 | Gain+Loss | LOC107985330 TP53BP2 CAPN2 FBXO28 LOC100287497 GTF2IP20 |
nsv832681 | Chr.1:223524706 - 223814989 on Build GRCh38 | Gain | CAPN8 TP53BP2 CAPN2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |