Assay Details
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4n111 | Chr.1:869076 - 914637 on Build GRCh38 | Duplication | LOC284600 FAM41C |
nsv1004962 | Chr.1:629241 - 915295 on Build GRCh38 | Gain | LOC100287934 MIR6723 LOC107984841 MTND1P23 LINC00115 OR4F16 LOC100133331 FAM87B LOC100288069 LINC01128 LOC284600 MTND2P28 FAM41C |
nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss | LOC107985728 FAM132A LOC105378591 PRKCZ LOC100288175 CCNL2 LOC105378947 LINC01128 B3GALT6 LOC101928626 TMEM88B LOC729737 FAAP20 SCNN1D MIR6859-2 RNF223 ATAD3C PUSL1 NADK OR4F16 LOC284600 TAS1R3 ANKRD65 GABRD NOC2L ISG15 MIR6727 MIR429 MIR200B UBE2J2 TMEM240 LINC01342 LOC107984841 MTND1P23 CPSF3L SLC35E2B LOC102724312 MXRA8 PLEKHN1 LOC100133331 LOC100288069 FNDC10 AGRN SDF4 MIB2 PERM1 DVL1 CPTP LOC148413 OR4F5 LOC100132287 MIR1302-2 SAMD11 TTLL10-AS1 LOC100134822 MIR200A SLC35E2 MRPL20 LOC100130417 VWA1 SKI TNFRSF18 GNB1 HES4 LINC00115 FAM87B MIR6808 LOC100129534 MIR6726 LOC107985729 ACAP3 ATAD3B MMP23B TMEM52 LOC102725121 WASH7P C1orf159 SSU72 MMP23A LOC100287934 MIR6723 MORN1 DDX11L1 FAM138A AURKAIP1 MTND2P28 FAM41C LOC105378592 LOC105378949 TTLL10 LOC105378948 TNFRSF4 OR4F29 CFAP74 LOC100506504 CDK11A MIR6859-1 ATAD3A KLHL17 CDK11B CALML6 LOC105378589 |
nsv950451 | Chr.1:899421 - 1094520 on Build GRCh38 | Deletion | LOC107985728 AGRN LOC105378948 PERM1 NOC2L ISG15 LOC100288175 RNF223 HES4 KLHL17 SAMD11 PLEKHN1 C1orf159 LOC100130417 LOC284600 |
nsv509035 | Chr.1:891406 - 985724 on Build GRCh38 | Insertion | LOC107985728 KLHL17 SAMD11 PLEKHN1 PERM1 NOC2L LOC100130417 LOC284600 |
dgv1n67 | Chr.1:811028 - 914557 on Build GRCh38 | Gain | LINC00115 FAM87B LINC01128 LOC284600 FAM41C |
nsv1005935 | Chr.1:805477 - 915295 on Build GRCh38 | Gain | LINC00115 FAM87B LINC01128 LOC284600 FAM41C |
esv3584982 | Chr.1:837710 - 917284 on Build GRCh38 | Gain | LOC100130417 LINC01128 LOC284600 FAM41C |
nsv1009537 | Chr.1:841742 - 915295 on Build GRCh38 | Gain | LINC01128 LOC284600 FAM41C |
nsv428334 | Chr.1:874371 - 1220569 on Build GRCh38 | Gain | LOC107985728 TNFRSF18 NOC2L ISG15 MIR429 MIR200B LOC100288175 HES4 LINC01342 PLEKHN1 FAM41C AGRN SDF4 TTLL10 LOC105378948 TNFRSF4 PERM1 RNF223 KLHL17 SAMD11 TTLL10-AS1 MIR200A C1orf159 LOC100130417 LOC284600 |
nsv10161 | Chr.1:776731 - 1777210 on Build GRCh38 | Gain+Loss | LOC107985728 TNFRSF18 FAM132A LOC100288175 HES4 LINC00115 CCNL2 FAM87B MIR6808 LINC01128 B3GALT6 MIR6726 TMEM88B LOC107985729 ACAP3 ATAD3B SCNN1D RNF223 ATAD3C PUSL1 NADK MMP23B C1orf159 SSU72 LOC284600 MMP23A TAS1R3 LOC100287934 ANKRD65 NOC2L ISG15 MIR6727 MIR429 MIR200B UBE2J2 TMEM240 LINC01342 CPSF3L SLC35E2B LOC102724312 MXRA8 PLEKHN1 LOC100288069 AURKAIP1 FNDC10 FAM41C AGRN SDF4 MIB2 TTLL10 LOC105378948 TNFRSF4 PERM1 CDK11A DVL1 CPTP LOC148413 ATAD3A KLHL17 SAMD11 CDK11B TTLL10-AS1 MIR200A SLC35E2 MRPL20 LOC100130417 VWA1 |
dgv5n100 | Chr.1:585989 - 1114424 on Build GRCh38 | Gain | LOC107985728 LOC100287934 MIR6723 NOC2L ISG15 LOC100288175 HES4 LOC107984841 MTND1P23 LINC00115 LOC105378947 PLEKHN1 LOC100133331 FAM87B LOC100288069 LINC01128 MTND2P28 FAM41C AGRN LOC101928626 LOC105378948 PERM1 RNF223 OR4F16 KLHL17 SAMD11 C1orf159 LOC100130417 LOC284600 |
nsv517709 | Chr.1:817186 - 1275912 on Build GRCh38 | Gain+Loss | LOC107985728 TNFRSF18 FAM132A NOC2L ISG15 MIR429 MIR200B LOC100288175 UBE2J2 HES4 LINC01342 LINC00115 PLEKHN1 FAM87B LINC01128 FAM41C B3GALT6 AGRN SDF4 TTLL10 LOC105378948 TNFRSF4 PERM1 RNF223 KLHL17 SAMD11 TTLL10-AS1 MIR200A C1orf159 LOC100130417 LOC284600 |
dgv1n111 | Chr.1:690090 - 939522 on Build GRCh38 | Duplication | LOC107985728 LOC100287934 LOC107984841 LINC00115 OR4F16 SAMD11 LOC100133331 FAM87B LOC100288069 LOC100130417 LINC01128 LOC284600 FAM41C |
dgv3n111 | Chr.1:783283 - 939522 on Build GRCh38 | Duplication | LOC107985728 LOC100287934 LINC00115 SAMD11 FAM87B LOC100130417 LINC01128 LOC284600 FAM41C |
dgv2n67 | Chr.1:877618 - 1426500 on Build GRCh38 | Gain | LOC107985728 ANKRD65 TNFRSF18 FAM132A NOC2L ISG15 MIR6727 MIR429 MIR200B LOC100288175 UBE2J2 HES4 LINC01342 CPSF3L CCNL2 MXRA8 PLEKHN1 AURKAIP1 MIR6808 B3GALT6 AGRN SDF4 TTLL10 LOC105378948 MIR6726 TMEM88B TNFRSF4 PERM1 ACAP3 DVL1 CPTP SCNN1D LOC148413 RNF223 PUSL1 KLHL17 SAMD11 TTLL10-AS1 MIR200A C1orf159 MRPL20 LOC100130417 LOC284600 TAS1R3 |
nsv832980 | Chr.1:848279 - 1007037 on Build GRCh38 | Gain | LOC107985728 KLHL17 SAMD11 PLEKHN1 PERM1 NOC2L LOC100130417 LINC01128 HES4 LOC284600 FAM41C |
esv3584976 | Chr.1:733250 - 914824 on Build GRCh38 | Gain | LOC100287934 LINC00115 FAM87B LOC100288069 LINC01128 LOC284600 LOC107984841 FAM41C |