Assay Details
Target Gene Details
Entrez Gene ID: | 100130197 |
Gene Name: | EFCAB14 antisense RNA 1 |
Gene Aliases: |
KIAA0494-AS1 |
Location: |
Chr.1:46674036-46692098 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EFCAB14-AS1 | NR_038827.1 | |||
NR_038828.1 | ||||
BE504902.1 | ||||
BX113937.1 |