Assay Details
Target Gene Details
Entrez Gene ID: | 10420 |
Gene Name: | testis-specific kinase 2 |
Gene Aliases: |
- |
Location: |
Chr.1:45343883-45491342 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TESK2 | NM_001320800.1 | NP_001307729.1 | ||
NM_007170.2 | NP_009101.2 | |||
XM_005270355.3 | XP_005270412.1 | |||
XM_006710287.2 | XP_006710350.1 | |||
XM_011540497.1 | XP_011538799.1 | |||
XM_011540500.2 | XP_011538802.1 | |||
XM_017000047.1 | XP_016855536.1 | |||
XM_017000048.1 | XP_016855537.1 | |||
XM_017000049.1 | XP_016855538.1 | |||
AB057597.1 | BAB62909.1 | |||
AJ132545.1 | CAB41970.1 | |||
AK027573.1 | ||||
AK294172.1 | ||||
BC033085.1 | AAH33085.1 | |||
BX648441.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2750837 | Chr.1:45330443 - 45442591 on Build GRCh38 | Loss | TOE1 MUTYH TESK2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |