Assay Details
Target Gene Details
Entrez Gene ID: | 100129269 |
Gene Name: | long intergenic non-protein coding RNA 1160 |
Gene Aliases: |
- |
Location: |
Chr.1:111599007-111608318 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01160 | NR_034126.1 | |||
AL553535.3 |
Target Gene Details
Entrez Gene ID: | 5906 |
Gene Name: | RAP1A, member of RAS oncogene family |
Gene Aliases: |
C21KG, G-22K, KREV-1, KREV1, RAP1, SMGP21 |
Location: |
Chr.1:111542223-111716695 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAP1A | XM_017001964.1 | XP_016857453.1 | ||
M22995.1 | AAA36150.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)