Assay Details
Target Gene Details
Entrez Gene ID: | 65260 |
Gene Name: | cytochrome c oxidase assembly factor 7 (putative) |
Gene Aliases: |
C1orf163, RESA1, SELRC1 |
Location: |
Chr.1:52686342-52698366 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COA7 | NM_023077.2 | NP_075565.2 | ||
AK022501.1 | BAB14063.1 | |||
AK225041.1 | ||||
AK225087.1 | ||||
BC015313.1 | AAH15313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain | LOC105378732 SLC1A7 PODN MIR1273F ECHDC2 MAGOH LOC100507564 ZYG11A LRP8 MIR5095 CPT2 MIR1273G ZYG11B SCP2 FAM159A COA7 C1orf123 |
esv2758934 | Chr.1:52660343 - 52823509 on Build GRCh38 | Loss | ZYG11B FAM159A COA7 |